siRNA Details
Gene Symbol: Ube4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207610.1 | NP_997493.1 | 15 | 2508 | ||
XM_006242938.2 | XP_006243000.1 | 16 | 2722 | |||
GenBank mRNA | AY325198.1 | - | 15 | 2460 | ||
BC061761.1 | - | 15 | 2508 | 5621078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
