siRNA Details
Gene Symbol: Bco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127712.1 | NP_001121184.1 | 7 | 961 | ||
GenBank mRNA | BC133725.1 | - | 7 | 961 | 7100438 | |
DQ083174.1 | - | 5 | 625 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127712.1 | NP_001121184.1 | 7 | 961 | ||
GenBank mRNA | BC133725.1 | - | 7 | 961 | 7100438 | |
DQ083174.1 | - | 5 | 625 |