siRNA Details
Gene Symbol: Ppp2r1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025418.1 | NP_001020589.1 | 13 | 1584 | ||
XM_006243032.2 | XP_006243094.1 | 15 | 6694 | |||
XM_006243033.2 | XP_006243095.1 | 14 | 6251 | |||
XM_006243034.2 | XP_006243096.1 | 15 | 6699 | |||
XM_008766260.1 | XP_008764482.1 | 16 | 5312 | |||
XM_008766261.1 | XP_008764483.1 | 16 | 5679 | |||
GenBank mRNA | BC098007.1 | - | 13 | 1584 | 7453957 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
