siRNA Details
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013176.1 | NP_001013194.1 | 3 | 385 | ||
XM_006243035.2 | XP_006243097.1 | 4 | 943 | |||
XM_006243036.2 | XP_006243098.1 | 3 | 343 | |||
XM_006243038.2 | XP_006243100.1 | 3 | 240 | |||
XM_008766262.1 | XP_008764484.1 | 3 | 377 | |||
GenBank mRNA | AY960132.1 | - | 2 | 190 | ||
BC088755.1 | - | 3 | 385 | 7307514 |
