siRNA Details
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013176.1 | NP_001013194.1 | 2, 3 | 369 | ||
XM_006243035.2 | XP_006243097.1 | 3, 4 | 927 | |||
XM_006243036.2 | XP_006243098.1 | 2, 3 | 327 | |||
XM_006243038.2 | XP_006243100.1 | 2, 3 | 224 | |||
XM_008766262.1 | XP_008764484.1 | 2, 3 | 361 | |||
GenBank mRNA | AY960132.1 | - | 1, 2 | 174 | ||
BC088755.1 | - | 2, 3 | 369 | 7307514 |

More Information
Other Species Targeted

Gene Ontology Categories:
