siRNA Details
Gene Symbol: Colca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243057.2 | XP_006243119.1 | 4 | 455 | ||
XM_006243058.2 | XP_006243120.1 | 4 | 462 | |||
XM_008766354.1 | XP_008764576.1 | 4 | 568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
