siRNA Details
Gene Symbol: Zc3h12c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108146.1 | NP_001101616.1 | 3, 4 | 978 | ||
XM_006243073.2 | XP_006243135.1 | 3, 4 | 1198 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001162921, AK220416.
Gene Ontology Categories:
