siRNA Details
Gene Symbol: Snx33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127488.1 | NP_001120960.1 | 1 | 1443 | ||
XM_006243123.2 | XP_006243185.1 | 2 | 2272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
