siRNA Details
Gene Symbol: Snx33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127488.1 | NP_001120960.1 | 1 | 2355 | ||
XM_006243123.2 | XP_006243185.1 | 2 | 3184 |

More Information
Other Species Targeted

Gene Ontology Categories:
