siRNA Details
Gene Symbol: Rpp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012124.1 | NP_001012124.1 | 1 | 567 | ||
GenBank mRNA | BC087592.1 | - | 1 | 567 | 7309941 |

More Information
Other Species Targeted

Gene Ontology Categories:
