siRNA Details
Gene Symbol: Sema7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108153.1 | NP_001101623.1 | 14 | 1800 | ||
XM_008766273.1 | XP_008764495.1 | 14 | 1647 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sema7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108153.1 | NP_001101623.1 | 14 | 1800 | ||
XM_008766273.1 | XP_008764495.1 | 14 | 1647 |