siRNA Details
Gene Symbol: Cd276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182824.2 | NP_877976.1 | 3 | 342 | ||
GenBank mRNA | AY190319.1 | - | 3 | 345 | ||
BC100064.1 | - | 3 | 342 | 7104832 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133983, DQ832276, BC019436, BC056608, AY190318, and others.
Gene Ontology Categories:
