siRNA Details
Gene Symbol: LOC103693110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766377.1 | XP_008764599.1 | 6 | 1284 | ||
XM_008766378.1 | XP_008764600.1 | 6 | 1181 | |||
XM_008766379.1 | XP_008764601.1 | 6 | 1130 |
