siRNA Details
Gene Symbol: Dpp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108159.1 | NP_001101629.1 | 19 | 2709 | ||
XM_006243252.2 | XP_006243314.1 | 19 | 2674 | |||
XM_006243253.2 | XP_006243315.1 | 19 | 2757 | |||
XM_006243254.2 | XP_006243316.1 | 19 | 2653 | |||
XM_006243255.2 | XP_006243317.1 | 18 | 2540 |
