siRNA Details
Gene Symbol: Rasl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108162.1 | NP_001101632.1 | 2, 3 | 257 | ||
XM_006243260.2 | XP_006243322.1 | 2, 3 | 265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
