siRNA Details
Gene Symbol: Cgnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108164.1 | NP_001101634.1 | 15 | 3475 | ||
XM_006243355.1 | XP_006243417.1 | 15 | 3419 | |||
XM_008766286.1 | XP_008764508.1 | 15 | 3820 | |||
XM_008766287.1 | XP_008764509.1 | 15 | 3475 |

More Information
Other Species Targeted

Gene Ontology Categories:
