siRNA Details
Gene Symbol: Zfp280d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108165.2 | NP_001101635.2 | 4 | 308 | ||
XM_006243356.2 | XP_006243418.1 | 3 | 275 | |||
XM_006243357.2 | XP_006243419.1 | 4 | 271 | |||
XM_008766288.1 | XP_008764510.1 | 3 | 258 | |||
XM_008766289.1 | XP_008764511.1 | 5 | 451 | |||
XM_008766290.1 | XP_008764512.1 | 3 | 258 | |||
GenBank mRNA | FQ222707.1 | - | 5 | 456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
