siRNA Details
Gene Symbol: Ttk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108172.1 | NP_001101642.1 | 9 | 1037 | ||
XM_006243460.2 | XP_006243522.1 | 10 | 1133 | |||
XM_006243461.2 | XP_006243523.1 | 10 | 1133 | |||
XM_006243462.2 | XP_006243524.1 | 9 | 1032 | |||
XM_006243463.2 | XP_006243525.1 | 9 | 1074 | |||
XM_006243464.2 | XP_006243526.1 | 10 | 1175 | |||
XM_006243465.2 | XP_006243527.1 | 9 | 1062 | |||
XM_006243466.2 | XP_006243528.1 | 9 | 1510 |
