siRNA Details
Gene Symbol: Snx14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108174.1 | NP_001101644.1 | 2 | 352 | ||
GenBank mRNA | BC160909.1 | - | 2 | 311 | 7455732 |

More Information
Other Species Targeted

Gene Ontology Categories:
