siRNA Details
Gene Symbol: U2surp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729954.4 | XP_002730000.1 | 14, 15 | 1411 | ||
XM_006243554.1 | XP_006243616.1 | 15, 16 | 1556 | |||
XM_236501.7 | XP_236501.3 | 15, 16 | 1597 | |||
GenBank mRNA | FQ226547.1 | - | 15, 16 | 1565 |
