siRNA Details
Gene Symbol: Vsig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037784.1 | NP_001032873.1 | 4 | 667 | ||
XM_006234243.2 | XP_006234305.1 | 5 | 2990 | |||
XM_006234244.2 | XP_006234306.1 | 4 | 2881 | |||
XM_006234245.2 | XP_006234307.1 | 3 | 3318 | |||
XM_006234246.2 | XP_006234308.1 | 3 | 1569 | |||
GenBank mRNA | BC098943.1 | - | 4 | 667 | 7456980 |
