siRNA Details
Gene Symbol: Lysmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009698.1 | NP_001009698.1 | 3 | 528 | ||
GenBank mRNA | BC088262.1 | - | 3 | 528 | 7376577 | |
FQ225025.1 | - | 2 | 246 | |||
FQ225894.1 | - | 2 | 246 | |||
FQ227395.1 | - | 2 | 246 | |||
FQ233176.1 | - | 2 | 246 | |||
FQ234752.1 | - | 2 | 246 |

More Information
Other Species Targeted

