siRNA Details
Gene Symbol: Zbtb38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012471.1 | NP_001012489.1 | 1 | 2604 | ||
XM_006243578.2 | XP_006243640.1 | 2 | 5380 | |||
XM_006243579.2 | XP_006243641.1 | 3 | 3119 | |||
XM_006243580.2 | XP_006243642.1 | 5 | 3245 | |||
XM_006243582.2 | XP_006243644.1 | 3 | 3048 | |||
XM_006243584.1 | XP_006243646.1 | 3 | 2702 | |||
XM_006243589.2 | XP_006243651.1 | 1 | 2543 | |||
XM_008766520.1 | XP_008764742.1 | 3 | 2930 | |||
XM_008766521.1 | XP_008764743.1 | 6 | 3404 | |||
XM_008766522.1 | XP_008764744.1 | 7 | 3415 | |||
XM_008766523.1 | XP_008764745.1 | 5 | 3300 | |||
XM_008766524.1 | XP_008764746.1 | 4 | 3179 | |||
GenBank mRNA | AY623002.1 | - | 1 | 2604 |

More Information
Other Species Targeted

Gene Ontology Categories:
