siRNA Details
Gene Symbol: Slc35g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012126.1 | NP_001012126.1 | 2 | 639 | ||
XM_006243630.2 | XP_006243692.1 | 2 | 807 | |||
GenBank mRNA | BC088758.1 | - | 2 | 639 | 7315776 |

More Information
Other Species Targeted

Gene Ontology Categories:
