siRNA Details
Gene Symbol: Msl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_236567.8 | XP_236567.5 | 2 | 1426 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Msl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_236567.8 | XP_236567.5 | 2 | 1426 |