siRNA Details
Gene Symbol: RGD1310507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047891.1 | NP_001041356.1 | 6 | 764 | ||
GenBank mRNA | AY325160.1 | - | 6 | 681 | ||
AY539912.1 | - | 6 | 764 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1310507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047891.1 | NP_001041356.1 | 6 | 764 | ||
GenBank mRNA | AY325160.1 | - | 6 | 681 | ||
AY539912.1 | - | 6 | 764 |