siRNA Details
Gene Symbol: Cdv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014097.1 | NP_001014119.1 | 2, 3 | 791 | ||
XM_006243659.2 | XP_006243721.1 | 2, 3 | 697 | |||
XM_006243660.2 | XP_006243722.1 | 2, 3 | 695 | |||
XM_008766529.1 | XP_008764751.1 | 2, 3 | 626 | |||
GenBank mRNA | BC083654.1 | - | 2, 3 | 791 | 7135192 |

More Information
Other Species Targeted

