siRNA Details
Gene Symbol: Dock3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750582.3 | XP_003750630.1 | 18 | 2179 | ||
XM_006243865.2 | XP_006243927.1 | 18 | 2179 | |||
XM_006243866.2 | XP_006243928.1 | 18 | 2179 | |||
XM_006243867.2 | XP_006243929.1 | 18 | 2179 | |||
XM_006243868.2 | XP_006243930.1 | 18 | 2179 |
