siRNA Details
Gene Symbol: Mapkapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012127.1 | NP_001012127.1 | 11 | 1359 | ||
XM_006243776.2 | XP_006243838.1 | 11 | 1449 | |||
XM_008766531.1 | XP_008764753.1 | 11 | 1458 | |||
GenBank mRNA | BC081974.1 | - | 11 | 1359 | 7126359 |
