siRNA Details
Gene Symbol: Setd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108189.1 | NP_001101659.1 | 3 | 3720 | ||
XM_008766632.1 | XP_008764854.1 | 2 | 4169 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Setd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108189.1 | NP_001101659.1 | 3 | 3720 | ||
XM_008766632.1 | XP_008764854.1 | 2 | 4169 |