siRNA Details
Gene Symbol: Als2cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244011.2 | XP_006244073.1 | 16 | 2790 | ||
XM_006244013.2 | XP_006244075.1 | 17 | 2622 | |||
XM_006244014.2 | XP_006244076.1 | 16 | 2821 | |||
XM_006244015.2 | XP_006244077.1 | 17 | 2877 | |||
XM_006244016.2 | XP_006244078.1 | 17 | 2874 | |||
XM_006244018.2 | XP_006244080.1 | 16 | 2997 | |||
XM_008766654.1 | XP_008764876.1 | 16 | 2824 | |||
XM_008766655.1 | XP_008764877.1 | 6 | 715 | |||
XM_236654.8 | XP_236654.6 | 16 | 3031 |
