siRNA Details
Gene Symbol: Ccrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108191.1 | NP_001101661.1 | 2 | 1548 | ||
XM_006243966.2 | XP_006244028.1 | 3 | 1321 | |||
GenBank mRNA | FQ225668.1 | - | 2 | 1481 |
