siRNA Details
Gene Symbol: Azi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025705.1 | NP_001020876.1 | 8 | 1488 | ||
GenBank mRNA | BC098680.1 | - | 8 | 1488 | 7384893 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Azi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025705.1 | NP_001020876.1 | 8 | 1488 | ||
GenBank mRNA | BC098680.1 | - | 8 | 1488 | 7384893 |