siRNA Details
Gene Symbol: Slc22a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126285.1 | NP_001119757.1 | 3, 4 | 660 | ||
XM_008766677.1 | XP_008764899.1 | 3, 4 | 666 | |||
XM_008766678.1 | XP_008764900.1 | 3, 4 | 666 | |||
GenBank mRNA | BC166511.1 | - | 3, 4 | 660 | 7128568 |
