siRNA Details
Gene Symbol: Exosc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100725.1 | NP_001094195.1 | 5, 6 | 506 | ||
GenBank mRNA | BC089995.1 | - | 4, 5 | 397 | ||
FQ226090.1 | - | 5, 6 | 511 |

More Information
Other Species Targeted

Gene Ontology Categories:
