siRNA Details
Gene Symbol: Lztfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024266.1 | NP_001019437.1 | 5 | 457 | ||
XM_006244191.1 | XP_006244253.1 | 5 | 601 | |||
GenBank mRNA | BC078913.1 | - | 5 | 553 | ||
BC092585.1 | - | 5 | 457 | 7127181 |

More Information
Other Species Targeted

Gene Ontology Categories:
