siRNA Details
Gene Symbol: Plin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_236783.6 | XP_236783.4 | 6 | 843 | ||
GenBank mRNA | FQ216473.1 | - | 2 | 183 | ||
FQ230643.1 | - | 6 | 780 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_236783.6 | XP_236783.4 | 6 | 843 | ||
GenBank mRNA | FQ216473.1 | - | 2 | 183 | ||
FQ230643.1 | - | 6 | 780 |