siRNA Details
Gene Symbol: Tfeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025707.1 | NP_001020878.1 | 3, 4 | 788 | ||
XM_006244440.2 | XP_006244502.1 | 4, 5 | 1717 | |||
XM_006244442.1 | XP_006244504.1 | 4, 5 | 707 | |||
XM_006244446.2 | XP_006244508.1 | 4, 5 | 1978 | |||
GenBank mRNA | BC099102.1 | - | 3, 4 | 788 | 7460021 |
