siRNA Details
Gene Symbol: Trerf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108199.1 | NP_001101669.1 | 7 | 2586 | ||
XM_006244452.2 | XP_006244514.1 | 12 | 3440 | |||
XM_006244453.2 | XP_006244515.1 | 12 | 3380 | |||
XM_008766863.1 | XP_008765085.1 | 11 | 3353 |

More Information
Other Species Targeted

Gene Ontology Categories:
