siRNA Details
Gene Symbol: Dlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108202.1 | NP_001101672.1 | 4 | 379 | ||
XM_006244523.2 | XP_006244585.1 | 4 | 346 | |||
XM_008766865.1 | XP_008765087.1 | 4 | 613 | |||
GenBank mRNA | BC169084.1 | - | 3 | 794 |

More Information
Other Species Targeted

Gene Ontology Categories:
