siRNA Details
Gene Symbol: Tjap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108203.1 | NP_001101673.1 | 11, 12 | 1044 | ||
XM_006244524.2 | XP_006244586.1 | 11, 12 | 1040 | |||
XM_006244525.2 | XP_006244587.1 | 11, 12 | 1037 | |||
XM_006244526.2 | XP_006244588.1 | 10, 11 | 910 | |||
XM_006244527.2 | XP_006244589.1 | 10, 11 | 942 | |||
XM_006244528.2 | XP_006244590.1 | 10, 11 | 939 | |||
XM_006244529.2 | XP_006244591.1 | 9, 10 | 812 | |||
XM_006244530.2 | XP_006244592.1 | 9, 10 | 809 | |||
XM_006244532.2 | XP_006244594.1 | 10, 11 | 937 | |||
XM_006244533.2 | XP_006244595.1 | 9, 10 | 807 | |||
XM_006244534.2 | XP_006244596.1 | 10, 11 | 1010 | |||
XM_006244535.2 | XP_006244597.1 | 8, 9 | 1305 | |||
GenBank mRNA | FQ221753.1 | - | 8, 9 | 1286 | ||
FQ221813.1 | - | 8, 9 | 1675 |
