siRNA Details
Gene Symbol: Nfkbie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199111.1 | NP_954542.1 | 2 | 645 | ||
GenBank mRNA | BC061795.1 | - | 2 | 645 | 5597840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
