siRNA Details
Gene Symbol: Enpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012744.1 | NP_001012762.1 | 2 | 652 | ||
XM_006244607.2 | XP_006244669.1 | 2 | 676 | |||
GenBank mRNA | BC090331.1 | - | 2 | 652 | 7300780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
