siRNA Details
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039028.2 | NP_001034117.2 | 7 | 781 | ||
GenBank mRNA | BC099152.1 | - | 2 | 375 | ||
BC166803.1 | - | 7 | 748 | 7938869 | ||
DY315273.1 | - | 4 | 394 | |||
FQ228979.1 | - | 7 | 749 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
