siRNA Details
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039028.2 | NP_001034117.2 | 7 | 817 | ||
GenBank mRNA | BC099152.1 | - | 2 | 411 | ||
BC166803.1 | - | 7 | 784 | 7938869 | ||
DY315273.1 | - | 4 | 430 | |||
FQ228979.1 | - | 7 | 785 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
