siRNA Details
Gene Symbol: Npas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108214.2 | NP_001101684.2 | 7 | 1096 | ||
XM_008767031.1 | XP_008765253.1 | 6 | 571 | |||
XM_008767032.1 | XP_008765254.1 | 7 | 1822 | |||
XM_008767033.1 | XP_008765255.1 | 7 | 1286 | |||
XM_008767034.1 | XP_008765256.1 | 7 | 2179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
