siRNA Details
Gene Symbol: Mfsd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108215.1 | NP_001101685.1 | 6 | 1383 | ||
XM_006244823.2 | XP_006244885.1 | 6 | 1375 | |||
XM_006244824.2 | XP_006244886.1 | 6 | 1519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
