siRNA Details
Gene Symbol: RGD1306941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108219.1 | NP_001101689.1 | 2 | 1000 | ||
GenBank mRNA | BC166506.1 | - | 2 | 721 |

More Information
Other Species Targeted

