siRNA Details
Gene Symbol: Tmem237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108220.1 | NP_001101690.1 | 9 | 1018 | ||
XM_006244975.2 | XP_006245037.1 | 10 | 1717 | |||
XM_006244976.2 | XP_006245038.1 | 10 | 1342 | |||
XM_006244977.2 | XP_006245039.1 | 10 | 1958 | |||
XM_008767098.1 | XP_008765320.1 | 11 | 1821 | |||
XM_008767099.1 | XP_008765321.1 | 11 | 1821 | |||
GenBank mRNA | BC166556.1 | - | 9 | 1672 |
