siRNA Details
Gene Symbol: Tmem237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108220.1 | NP_001101690.1 | 7 | 705 | ||
XM_006244975.2 | XP_006245037.1 | 8 | 1404 | |||
XM_006244976.2 | XP_006245038.1 | 8 | 1029 | |||
XM_006244977.2 | XP_006245039.1 | 8 | 1645 | |||
XM_008767098.1 | XP_008765320.1 | 8 | 1404 | |||
XM_008767099.1 | XP_008765321.1 | 8 | 1404 | |||
GenBank mRNA | BC166556.1 | - | 7 | 1359 |
